missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Endotoxin, Stem Cell Culture
Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation
71.00€ - 3205.00€
Specifications
For Use With (Application) | Mammalian Cell Culture |
---|---|
Formulation | Lipid-rich |
Molecular Weight (g/mol) | 66 |
Name | CellMaxx™ Bovine Albumin, Low Endotoxin |
pH Range | 6.5 to 7.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17308573
|
MP Biomedicals™
0219989610 |
10 g |
71.00€
10g |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
17358583
|
MP Biomedicals™
0219989625 |
25 g |
121.00€
25g |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
17318573
|
MP Biomedicals™
0219989650 |
50 g |
235.00€
50g |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
11401655
|
MP Biomedicals™
0219989680 |
100 g |
372.00€
100g |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
15702272
|
MP Biomedicals™
0219989690 |
500 g |
1680.00€
500g |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
11411655
|
MP Biomedicals™
0219989691 |
1000 g |
3205.00€
1kg |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Lipid-rich formulation can be substituted for Albumax™ II.
Physical characteristics:
- Form: Off white to brown powder
- Purity:≥98% (w/w)
- Total Protein: ≥96%
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Certifications: ISO9001, ISO13485, cGMP, EDQM
Specifications
Mammalian Cell Culture | |
66 | |
6.5 to 7.5 | |
Bovine | |
Powder |
Lipid-rich | |
CellMaxx™ Bovine Albumin, Low Endotoxin | |
Chromatography | |
4°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title