Learn More
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts
Choose these replacement parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
18.60€ - 399.00€
Specifications
For Use With (Equipment) | Owl™ A3-1 and A6 Wide Gel System |
---|---|
Product Line | Owl |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11952519
|
Thermo Scientific™
A3-1-EG |
68.20€
Pack of 2 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||||
12444537
|
Thermo Scientific™
A3-GK-1 |
18.60€
2 sets |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||||
12454537
|
Thermo Scientific™
A3-UVT-1 |
UVT gel tray with gasketed end gates |
399.00€
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
Description
Choose Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
The gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run utilizing a wall comb. Built in buffer exchange ports allows buffer recirculation on extended runs, if needed. The large number of comb slots gives you sample and resolving distance flexibility.Specifications
Owl™ A3-1 and A6 Wide Gel System | |
Owl |
Product Title
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts