missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase XIAP Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase XIAP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6891-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001158.2 | |
E3 ubiquitin-protein ligase XIAP Protein | |
Research Use Only | |
XIAP | |
Recombinant Protein | |
E. coli | |
MTFNSFEGSK TCVPADINKE EEFVEEFNRL KTFANFPSGS PVSASTLARA GFLYTGEGDT VRCFSCHAAV DRWQYGDSAV GRHRKVSPNC RFINGFYLEN SATQSTNSGI QNGQYKVENY LGSRDHFALD RPSETHADYL LRTGQVVDIS DTIYPRNPAM YSEEARLKSF QNWPDYAHLT PRELASAGLY YTGIGDQVQC FCCGGKLKNW EPCDRAWSEH RRHFPNCFFV LGRNLNIRSE SDAVSSDRNF PNSTNLPRNP SMADYEARIF TFGTWIYSVN KEQLARAGFY ALGEGDKVKC FHCGGGLTDW KPSEDPWEQH AKWYPGCKYL LEQKGQEYIN NIHLTHSLEE CLVRTTEKTP SLTRRIDDTI FQNPMVQEAI RMGFSFKDIK KIMEEKIQIS GSNYKSLEVL VADLVNAQKD SMQDESSQTS LQKEISTEEQ LRRLQEEKLC KICMDRNIAI VFVPCGHLVT CKQCAEAVDK CPMCYTVITF KQKIFMS | |
Greater than 90% as determined by SDS-PAGE. |
331 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase XIAP Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction