missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase XIAP Protein

A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase XIAP, was expressed in the hosts and tags indicated.

160.00€ - 2485.00€

Specifications

Accession Number NP_001158.2
Gene ID (Entrez) 331
Name E3 ubiquitin-protein ligase XIAP Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16046590
View Documents
enQuireBio™
QP6891-ec-10ug
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16086590
View Documents
enQuireBio™
QP6891-ec-50ug
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16036590
View Documents
enQuireBio™
QP6891-ec-100ug
100 μg
554.00€
100µg
Please to purchase this item. Need a web account? Register with us today!
16066590
View Documents
enQuireBio™
QP6891-ec-200ug
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16076590
View Documents
enQuireBio™
QP6891-ec-500ug
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16056590
View Documents
enQuireBio™
QP6891-ec-1mg
1 mg
2485.00€
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001158.2
E3 ubiquitin-protein ligase XIAP Protein
Not determined.
Full Length
Human
Tris-based buffer, 50% glycerol
331
Research Use Only
XIAP
Recombinant Protein
MTFNSFEGSK TCVPADINKE EEFVEEFNRL KTFANFPSGS PVSASTLARA GFLYTGEGDT VRCFSCHAAV DRWQYGDSAV GRHRKVSPNC RFINGFYLEN SATQSTNSGI QNGQYKVENY LGSRDHFALD RPSETHADYL LRTGQVVDIS DTIYPRNPAM YSEEARLKSF QNWPDYAHLT PRELASAGLY YTGIGDQVQC FCCGGKLKNW EPCDRAWSEH RRHFPNCFFV LGRNLNIRSE SDAVSSDRNF PNSTNLPRNP SMADYEARIF TFGTWIYSVN KEQLARAGFY ALGEGDKVKC FHCGGGLTDW KPSEDPWEQH AKWYPGCKYL LEQKGQEYIN NIHLTHSLEE CLVRTTEKTP SLTRRIDDTI FQNPMVQEAI RMGFSFKDIK KIMEEKIQIS GSNYKSLEVL VADLVNAQKD SMQDESSQTS LQKEISTEEQ LRRLQEEKLC KICMDRNIAI VFVPCGHLVT CKQCAEAVDK CPMCYTVITF KQKIFMS
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase XIAP Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.