missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PFDN5 / MM-1 Protein

A DNA sequence encoding the Homo sapiens (Human) PFDN5 / MM-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8367-ec-10ug

 View more versions of this product

Product Code. 16004261

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_002615.2
PFDN5 / MM-1 Protein
Research Use Only
PFDN5
Recombinant Protein
E. coli
AQSINITELN LPQLEMLKNQ LDQEVEFLST SIAQLKVVQT KYVEAKDCLN VLNKSNEGKE LLVPLTSSMY VPGKLHDVEH VLIDVGTGYY VEKTAEDAKD FFKRKIDFLT KQMEKIQPAL QEKHAMKQAV MEMMSQKIQQ LTALGAAQAT AKA
Greater than 90% as determined by SDS-PAGE.
5204
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers