missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PFDN5 / MM-1 Protein
A DNA sequence encoding the Homo sapiens (Human) PFDN5 / MM-1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_002615.2 |
---|---|
Gene ID (Entrez) | 5204 |
Name | PFDN5 / MM-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004261
|
enQuireBio™
QP8367-ec-10ug |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16044261
|
enQuireBio™
QP8367-ec-50ug |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16094251
|
enQuireBio™
QP8367-ec-100ug |
100 μg |
530.00€
100µg |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
16024261
|
enQuireBio™
QP8367-ec-200ug |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16034261
|
enQuireBio™
QP8367-ec-500ug |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16014261
|
enQuireBio™
QP8367-ec-1mg |
1 mg |
2385.00€
1mg |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
Specifications
NP_002615.2 | |
PFDN5 / MM-1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5204 | |
Research Use Only | |
PFDN5 | |
Recombinant Protein | |
E. coli | |
AQSINITELN LPQLEMLKNQ LDQEVEFLST SIAQLKVVQT KYVEAKDCLN VLNKSNEGKE LLVPLTSSMY VPGKLHDVEH VLIDVGTGYY VEKTAEDAKD FFKRKIDFLT KQMEKIQPAL QEKHAMKQAV MEMMSQKIQQ LTALGAAQAT AKA | |
Greater than 90% as determined by SDS-PAGE. |