missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSAP / Prosaposin Protein

A DNA sequence encoding the Homo sapiens (Human) PSAP / Prosaposin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6540-ec-10ug

 View more versions of this product

Product Code. 16084030

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001035930.1
PSAP / Prosaposin Protein
Research Use Only
PSAP
Recombinant Protein
E. coli
SDVYCEVCEF LVKEVTKLID NNKTEKEILD AFDKMCSKLP KSLSEECQEV VDTYGSSILS ILLEEVSPEL VCSMLHLCSG T
Greater than 90% as determined by SDS-PAGE.
5660
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers