missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSAP / Prosaposin Protein
A DNA sequence encoding the Homo sapiens (Human) PSAP / Prosaposin, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001035930.1 |
---|---|
Gene ID (Entrez) | 5660 |
Name | PSAP / Prosaposin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084030
|
enQuireBio™
QP6540-ec-10ug |
10 μg | N/A | ||||||
16024040
|
enQuireBio™
QP6540-ec-50ug |
50 μg | N/A | ||||||
16074030
|
enQuireBio™
QP6540-ec-100ug |
100 μg |
530.00€
100µg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16004040
|
enQuireBio™
QP6540-ec-200ug |
200 μg | N/A | ||||||
16014040
|
enQuireBio™
QP6540-ec-500ug |
500 μg | N/A | ||||||
16094030
|
enQuireBio™
QP6540-ec-1mg |
1 mg |
2385.00€
1mg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Specifications
NP_001035930.1 | |
PSAP / Prosaposin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
Tris-based buffer, 50% glycerol |
5660 | |
Research Use Only | |
PSAP | |
Recombinant Protein | |
SDVYCEVCEF LVKEVTKLID NNKTEKEILD AFDKMCSKLP KSLSEECQEV VDTYGSSILS ILLEEVSPEL VCSMLHLCSG T |