missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thyrotropin subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Thyrotropin subunit beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6844-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_000540.2 | |
Thyrotropin subunit beta Protein | |
Research Use Only | |
TSHB | |
Recombinant Protein | |
E. coli | |
FCIPTEYTMH IERRECAYCL TINTTICAGY CMTRDINGKL FLPKYALSQD VCTYRDFIYR TVEIPGCPLH VAPYFSYPVA LSCKCGKCNT DYSDCIHEAI KTNYCTKPQK SY | |
Greater than 90% as determined by SDS-PAGE. |
7252 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |