missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thyrotropin subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Thyrotropin subunit beta, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_000540.2 |
---|---|
Gene ID (Entrez) | 7252 |
Name | Thyrotropin subunit beta Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006240
|
enQuireBio™
QP6844-ec-10ug |
10 μg | N/A | ||||||
16046240
|
enQuireBio™
QP6844-ec-50ug |
50 μg | N/A | ||||||
16096230
|
enQuireBio™
QP6844-ec-100ug |
100 μg |
530.00€
100µg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16026240
|
enQuireBio™
QP6844-ec-200ug |
200 μg | N/A | ||||||
16036240
|
enQuireBio™
QP6844-ec-500ug |
500 μg | N/A | ||||||
16016240
|
enQuireBio™
QP6844-ec-1mg |
1 mg |
2385.00€
1mg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Specifications
NP_000540.2 | |
Thyrotropin subunit beta Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7252 | |
Research Use Only | |
TSHB | |
Recombinant Protein | |
E. coli | |
FCIPTEYTMH IERRECAYCL TINTTICAGY CMTRDINGKL FLPKYALSQD VCTYRDFIYR TVEIPGCPLH VAPYFSYPVA LSCKCGKCNT DYSDCIHEAI KTNYCTKPQK SY | |
Greater than 90% as determined by SDS-PAGE. |