missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tumor protein D53 Protein

A DNA sequence encoding the Homo sapiens (Human) Tumor protein D53, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7623-ec-1mg

 View more versions of this product

Product Code. 16019860

  • 2385.00€ / 1mg
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001003395.1
Tumor protein D53 Protein
Research Use Only
TPD52L1
Recombinant Protein
E. coli
MEAQAQGLLE TEPLQGTDED AVASADFSSM LSEEEKEELK AELVQLEDEI TTLRQVLSAK ERHLVEIKQK LGMNLMNELK QNFSKSWHDM QTTTAYKKTH ETLSHAGQKA TAAFSNVGTA ISKKFGDMSY SIRHSISMPA MRNSPTFKSF EERVETTVTS LKTKVGGTNP NGGSFEEVLS STAHASAQSL AGGSRRTKEE ELQC
Greater than 80% as determined by SDS-PAGE.
7164
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers