missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tumor protein D53 Protein
A DNA sequence encoding the Homo sapiens (Human) Tumor protein D53, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001003395.1 |
---|---|
Gene ID (Entrez) | 7164 |
Name | Tumor protein D53 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009860
|
enQuireBio™
QP7623-ec-10ug |
10 μg | N/A | ||||||
16049860
|
enQuireBio™
QP7623-ec-50ug |
50 μg | N/A | ||||||
16099850
|
enQuireBio™
QP7623-ec-100ug |
100 μg |
530.00€
100µg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16029860
|
enQuireBio™
QP7623-ec-200ug |
200 μg | N/A | ||||||
16039860
|
enQuireBio™
QP7623-ec-500ug |
500 μg | N/A | ||||||
16019860
|
enQuireBio™
QP7623-ec-1mg |
1 mg |
2385.00€
1mg |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Specifications
NP_001003395.1 | |
Tumor protein D53 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
7164 | |
Research Use Only | |
TPD52L1 | |
Recombinant Protein | |
E. coli | |
MEAQAQGLLE TEPLQGTDED AVASADFSSM LSEEEKEELK AELVQLEDEI TTLRQVLSAK ERHLVEIKQK LGMNLMNELK QNFSKSWHDM QTTTAYKKTH ETLSHAGQKA TAAFSNVGTA ISKKFGDMSY SIRHSISMPA MRNSPTFKSF EERVETTVTS LKTKVGGTNP NGGSFEEVLS STAHASAQSL AGGSRRTKEE ELQC | |
Greater than 80% as determined by SDS-PAGE. |