missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse KIM-1 / TIM1 Protein
A DNA sequence encoding the Mus musculus (Mouse) KIM-1 / TIM1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8888-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001160103.1 | |
KIM-1 / TIM1 Protein | |
Research Use Only | |
Havcr1 | |
Recombinant Protein | |
E. coli | |
YVEVKGVVGH PVTLPCTYST YRGITTTCWG RGQCPSSACQ NTLIWTNGHR VTYQKSSRYN LKGHISEGDV SLTIENSVES DSGLYCCRVE IPGWFNDQKV TFSLQVKPEI PTRPPTRPTT TRPTATGRPT TISTRSTHVP TSIRVSTSTP PTSTHTWTHK PEPTTFCPHE TTAEVTGIPS HTPTDWNGTV TSSGDTWSNH TEAIPPGKPQ KNPTKG | |
Greater than 90% as determined by SDS-PAGE. |
171283 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |