missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse KIM-1 / TIM1 Protein

A DNA sequence encoding the Mus musculus (Mouse) KIM-1 / TIM1, was expressed in the hosts and tags indicated.

245.00€ - 3405.00€

Specifications

Accession Number NP_001160103.1
Gene ID (Entrez) 171283
Name KIM-1 / TIM1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16007001
View Documents Promotion Details
enQuireBio™
QP8888-ec-10ug
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16047001
View Documents Promotion Details
enQuireBio™
QP8888-ec-50ug
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16096991
View Documents Promotion Details
enQuireBio™
QP8888-ec-100ug
100 μg
770.00€
100µg
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
16027001
View Documents Promotion Details
enQuireBio™
QP8888-ec-200ug
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16037001
View Documents Promotion Details
enQuireBio™
QP8888-ec-500ug
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16017001
View Documents Promotion Details
enQuireBio™
QP8888-ec-1mg
1 mg
3405.00€
1mg
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

NP_001160103.1
KIM-1 / TIM1 Protein
Not determined.
Extracellular Domain
Mouse
Tris-based buffer, 50% glycerol
171283
Research Use Only
Havcr1
Recombinant Protein
YVEVKGVVGH PVTLPCTYST YRGITTTCWG RGQCPSSACQ NTLIWTNGHR VTYQKSSRYN LKGHISEGDV SLTIENSVES DSGLYCCRVE IPGWFNDQKV TFSLQVKPEI PTRPPTRPTT TRPTATGRPT TISTRSTHVP TSIRVSTSTP PTSTHTWTHK PEPTTFCPHE TTAEVTGIPS HTPTDWNGTV TSSGDTWSNH TEAIPPGKPQ KNPTKG
Videos
SDS
Documents

Documents

Product Certifications