Learn More
R&D Systems™ Recombinant Mouse LRP-5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 7344-LR-025
 View more versions of this product
Product Code. 18364747
  • 456.00€ / 25µg
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Recombinant Mouse LRP-5 Protein is derived from HEK293. The Recombinant Mouse LRP-5 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
152.1 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
16973
LRP-5
Human embryonic kidney cell,HEK293-derived mouse LRP-5 protein Met1-Ser1383,with a C-terminal 6-His tag
Unconjugated
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.