missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse LRP-5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
456.00€
Specifications
Gene ID (Entrez) | 16973 |
---|---|
Molecular Weight (g/mol) | 152.1 kDa |
Name | LRP-5 |
Quantity | 25 μg |
Source | Human embryonic kidney cell,HEK293-derived mouse LRP-5 protein Met1-Ser1383,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18364747
|
R&D Systems™
7344-LR-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
456.00€
25µg |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
18107460
|
R&D Systems™
7344-LR-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
456.00€
25µg |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
The Recombinant Mouse LRP-5 Protein is derived from HEK293. The Recombinant Mouse LRP-5 Protein has been validated for the following applications: Bioactivity.Specifications
16973 | |
LRP-5 | |
Human embryonic kidney cell,HEK293-derived mouse LRP-5 protein Met1-Ser1383,with a C-terminal 6-His tag | |
Unconjugated | |
85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
152.1 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title