missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat TIMP-1 / TIMP1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) TIMP-1 / TIMP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8876-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_446271.1 | |
TIMP-1 / TIMP1 Protein | |
Research Use Only | |
Timp1 | |
Recombinant Protein | |
E. coli | |
CSCAPTHPQT AFCNSDLVIR AKFMGSPEII ETTLYQRYEI KMTKMLKGFD AVGNATGFRF AYTPAMESLC GYVHKSQNRS EEFLIAGRLR NGNLHITACS FLVPWHNLSP AQQKAFVKTY SAGCGVCTVF PCSAIPCKLE SDSHCLWTDQ ILMGSEKGYQ SDHFACLPRN PDLCTWQYLG VSMTRSLPLA KAEA | |
Greater than 90% as determined by SDS-PAGE. |
116510 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |