missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat TIMP-1 / TIMP1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) TIMP-1 / TIMP1, was expressed in the hosts and tags indicated.

245.00€ - 3405.00€

Specifications

Accession Number NP_446271.1
Gene ID (Entrez) 116510
Name TIMP-1 / TIMP1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16086921
View Documents Promotion Details
enQuireBio™
QP8876-ec-10ug
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16026931
View Documents Promotion Details
enQuireBio™
QP8876-ec-50ug
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16076921
View Documents Promotion Details
enQuireBio™
QP8876-ec-100ug
100 μg
770.00€
100µg
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
16006931
View Documents Promotion Details
enQuireBio™
QP8876-ec-200ug
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16016931
View Documents Promotion Details
enQuireBio™
QP8876-ec-500ug
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16096921
View Documents Promotion Details
enQuireBio™
QP8876-ec-1mg
1 mg
3405.00€
1mg
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

NP_446271.1
TIMP-1 / TIMP1 Protein
Not determined.
Full Length
Rat
Tris-based buffer, 50% glycerol
116510
Research Use Only
Timp1
Recombinant Protein
CSCAPTHPQT AFCNSDLVIR AKFMGSPEII ETTLYQRYEI KMTKMLKGFD AVGNATGFRF AYTPAMESLC GYVHKSQNRS EEFLIAGRLR NGNLHITACS FLVPWHNLSP AQQKAFVKTY SAGCGVCTVF PCSAIPCKLE SDSHCLWTDQ ILMGSEKGYQ SDHFACLPRN PDLCTWQYLG VSMTRSLPLA KAEA
Videos
SDS
Documents

Documents

Product Certifications