Learn More
Corning™ rLaminin-521 (Human)
Recombinant heterotrimer protein composed of α5, β2, and γ1 chains
69.20€ - 3948.30€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15689176
|
Corning™
354221 |
100 ÎĽg |
69.20€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15930151
|
Corning™
354223 |
5 mg | N/A | ||||||
15618068
|
Corning™
354223 |
5 mg | N/A | ||||||
15608068
|
Corning™
354222 |
1 mg | N/A | ||||||
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning rLaminin-521 is the next generation pluripotent stem cell (PSC) surface that enables researchers to streamline culture workflow, lower contamination risk, and scale-up efficiently. Corning has partnered with BioLamina for the supply of recombinant human Laminin 521 (rLaminin-521), a biocompatible, full length Laminin that enables feeder-free, clump-free, single-cell passaging of PSCs without ROCK-inhibitor.
Specifications
rLaminin-521, Human | |
Human | |
Recombinant |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.