Learn More
Corning™ rLaminin-521 (Human)
Recombinant heterotrimer protein composed of α5, β2, and γ1 chains
Brand:  Corning™ 354222
 View more versions of this product
Product Code. 15608068
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning rLaminin-521 is the next generation pluripotent stem cell (PSC) surface that enables researchers to streamline culture workflow, lower contamination risk, and scale-up efficiently. Corning has partnered with BioLamina for the supply of recombinant human Laminin 521 (rLaminin-521), a biocompatible, full length Laminin that enables feeder-free, clump-free, single-cell passaging of PSCs without ROCK-inhibitor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
rLaminin-521, Human
1 mg
Human
Recombinant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ rLaminin-521 (Human) > rLaminin-521, Human, 1 mg; 100 mg