missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SUB1 (Human) Recombinant Protein

Human SUB1 partial ORF ( AAH09610.1, 32 a.a. - 124 a.a.) recombinant protein with GST-tag at N-terminal.

344.00€ - 521.00€

Specifications

Accession Number AAH09610.1
Gene ID (Entrez) 10923
Name SUB1 homolog (S. cerevisiae)
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16185862
View Documents
Abnova™
H00010923-Q01.10ug
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
16195862
View Documents
Abnova™
H00010923-Q01.25ug
25 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description

Description

  • Sequence: APEKPVKKQKTGETSRALSSSKQSSSSRDDNMFQIGKMRYVSVRDFKGKVLIDIREYWMDPEGEMKPGRKGISLNPEQWSQLKEQISDIDDAV
Specifications

Specifications

AAH09610.1
SUB1 homolog (S. cerevisiae)
125% SDS-PAGE Stained with Coomassie Blue
MGC102747, P15, PC4, p14
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
10923
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SUB1
GST
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.