Learn More
3M™ Versaflo™ BT-Series Breathing Tube Covers
Brand:  3M™ 7000103964
17.99 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15709057
  • 29.75€ / Pack of 5
Estimated Shipment: 12-05-2025
Specifications
Disposable Cover Plastic Pipes
Versaflo BT-Series breathing tubes
5/Pk.
Videos
  • 3M - Powered and supplied air products
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M Powered and supplied air products
    Product Title
    3M™ Versaflo™ BT-Series Breathing Tube Covers > Disposable