Learn More
Thermo Scientific™ Cryoware Permanent Markers and Pens
Identify samples and storage boxes permanently with these cryoware permanent markers and pens.
Brand: Thermo Scientific™ 339993
40.84 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.
- Nunc™ and Nalgene™ cryoware markers come in a variety of colors for permanent handwritten identification of samples
- Use these markers by hand, or for higher throughput use them in the ID Scribe
Specifications
Assorted (Red, Blue, Green, Black) | |
Nunc Cryo Pen Permanent Marker Set | |
100 | |
4 |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
167972 | Certificate of Compliance | 03/12/2024 | 339993 |
182101 | Certificate of Compliance | 03/12/2024 | 339993 |
180159 | Certificate of Compliance | 03/12/2024 | 339993 |
164827 | Certificate of Compliance | 03/12/2024 | 339993 |
179042 | Certificate of Compliance | 03/12/2024 | 339993 |