missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Microplate Lids
GREENER_CHOICE

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Brand:  Thermo Scientific™ 250003

 View more versions of this product

Product Code. 11817184

  • 655.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
384/1536
5 in.
No
60
5 x 3.35 in. (127 x 85 mm)
3.35 in.
Universal Lid
None
All well shapes
No
5 x 3.35 x 0.33 in. (127 x 85 x 8.34 mm)
Non-sterile
8.34 mm
127 mm
0.33 in.
20
No
85 mm
Nunc
-20°C to +80°C
All Microplates
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Schematics & Diagrams

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1420332 Certificate TypeCertificate of Compliance Date28/01/2025 Catalog Number250003
Lot Number1420331 Certificate TypeCertificate of Compliance Date28/01/2025 Catalog Number250003
Lot Number1418068 Certificate TypeCertificate of Compliance Date14/12/2024 Catalog Number250003
Lot Number1336947 Certificate TypeCertificate of Compliance Date11/12/2024 Catalog Number250003
Lot Number1416602 Certificate TypeCertificate of Compliance Date19/11/2024 Catalog Number250003
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ Microplate Lids > Universal Lid for 384 and 1536 well Nunc plate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.